Assay Details
| Gene Symbol: Yy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK137406.1 | - | 1951 | 95 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Downstream |
solute carrier family 25 (mitochondrial carrier, palmitoylcarnitine transporter), member 29 |
Slc25a29 |
C030003J19Rik, CACL, mCACL |
5.7kb | Sense | Chr. 12 - 108825878 - 108835876 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Yy1
Genomic Map