Assay Details
| Gene Symbol: Zfp229 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164676.1 | NP_001158148.1 | 6 | 3331 | 99 | |
| GenBank mRNA | AK137438.1 | - | 1 | 1043 | 99 | |
| BC043476.1 | - | 3 | 2922 | 99 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 760 | Zfp760 |
9430018I06, BC029103 |
-21.1kb | Sense | Chr. 17 - 21707741 - 21725636 on NCBI Build GRCm38 |
| Downstream | zinc finger protein 820 | Zfp820 | 2610036F08Rik | 67.9kb | Anti-sense | Chr. 17 - 21816876 - 21846022 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Zfp229
Genomic Map