Assay Details
| Gene Symbol: A230045G11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK133970.1 | - | 1 - 2 | 208 | 89 | |
| AK138253.1 | - | 1 - 2 | 164 | 89 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
phosphatidylserine decarboxylase, pseudogene 2 |
Pisd-ps2 | D030013I16Rik | -30.0kb | Sense | Chr. 17 - 3064318 - 3084183 on NCBI Build GRCm38 |
| Downstream | SR-related CTD-associated factor 8 | Scaf8 |
A630086M08Rik, A930036P18Rik, AA408306, AI447644, AI448652, AU015035, Rbm16, mKIAA1116 |
222.0 | Anti-sense | Chr. 17 - 3114972 - 3198859 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

A230045G11Rik
Genomic Map