Array Name |
Probe IDs |
Targets |
Target Type |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
795725 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
381895 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
310123 |
AK077863.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
310123 |
AK131889.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
310123 |
AK167380.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
310123 |
AK170311.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
310123 |
BC024135.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
310123 |
BC051055.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
310123 |
BC088736.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
310123 |
BC132168.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
310123 |
BC132170.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
310123 |
NM_172582.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
52795 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
49465 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2179060 |
AK138389.1 |
mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05488397 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05488417 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23497998 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23497997 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23497988 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23497995 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23497986 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23497990 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23498001 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23497982 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23497992 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23497999 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23497994 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23497981 |
- |
- |