Assay Details
| Gene Symbol: Gm1715 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045333.1 | 2 - 3 | 364 | 86 | ||
| GenBank mRNA | AK138605.1 | - | 2 - 3 | 365 | 86 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | expressed sequence AI593442 | AI593442 | E130102D23 | -36.6kb | Anti-sense | Chr. 9 - 52673032 - 52680064 on NCBI Build GRCm38 |
| Downstream | DEAD (Asp-Glu-Ala-Asp) box polypeptide 10 | Ddx10 |
4632415A01Rik, AI646054 |
322.3kb | Anti-sense | Chr. 9 - 53098454 - 53248170 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm1715
Genomic Map