Assay Details
Gene Symbol: Gm1715 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045333.1 | 4 - 5 | 674 | 79 | ||
GenBank mRNA | AK138605.1 | - | 4 - 5 | 675 | 79 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | expressed sequence AI593442 | AI593442 | E130102D23 | -102.6kb | Anti-sense | Chr. 9 - 52673032 - 52680064 on NCBI Build GRCm38 |
Downstream | DEAD (Asp-Glu-Ala-Asp) box polypeptide 10 | Ddx10 |
4632415A01Rik, AI646054 |
299.7kb | Anti-sense | Chr. 9 - 53098454 - 53248170 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1955422 | AK138605.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 266497 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 649106 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 908327 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 210293 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 634263 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 406975 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 829532 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 761438 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 743347 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 506747 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 902059 | AK009895.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 902059 | AK011657.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 902059 | AK078891.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 902059 | AK088122.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 902059 | AK134177.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 902059 | AK135298.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 902059 | AK141440.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 902059 | AK145089.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 902059 | AK145600.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 902059 | AK145715.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 902059 | AK146010.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 902059 | AK146253.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 902059 | AK146610.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 902059 | AK152411.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 902059 | AK152704.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 902059 | AK152779.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 902059 | AK153073.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 902059 | AK153209.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 902059 | AK154046.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 902059 | BC068111.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 902059 | BC071196.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 902059 | BC083316.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 902059 | NM_013663.4 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 902059 | X53824.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 902059 | XM_001479186.1 | refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 902059 | XM_001480979.1 | refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 514404 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 211759 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 107848 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 260215 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 432349 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 666599 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 758102 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 426294 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 960229 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 61865 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 254369 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 400574 | AK034901.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 791628 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 238475 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 407864 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 731486 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 391388 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 744667 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 754068 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 73429 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 280178 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 163917 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 967870 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 245012 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 503831 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 972270 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 546533 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 378100 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 693399 | AK039973.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 637930 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 899613 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 746247 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 108739 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 168738 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 152073 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 69501 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 16892 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 30211 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 84028 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 60545 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P146649 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1955422 | AK138605.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P146649 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06790407 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06790601 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06790491 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01933926 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01933852 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06790424 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01933892 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06790577 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06790445 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06790537 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06790421 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01933984 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06790421 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06790445 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P146649 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01933984 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06790537 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516832 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516828 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516711 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516792 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07813009 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516818 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516686 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516746 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516780 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516776 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516748 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516753 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08126417 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516907 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516909 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516674 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516671 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516799 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516708 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516900 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516754 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07813028 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516678 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516912 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516806 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516700 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516679 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07924278 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516789 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07479951 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516798 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516810 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516696 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516683 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07556167 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516903 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516750 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516677 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516809 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07805544 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516706 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516786 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516820 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516922 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516687 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07580188 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516744 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07812991 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516824 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516751 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516705 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516784 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516825 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516783 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516703 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516741 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516902 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516826 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07827114 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516673 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516791 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516777 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516910 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516916 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516816 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516772 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07645991 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07852773 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516918 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516714 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516781 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516698 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516712 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516914 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516822 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516681 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516685 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516794 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516829 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516701 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516782 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516779 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516796 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516775 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516803 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26516830 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08095110 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07812989 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Clariom S Array | PSR0900004531.mm.1 | NR_045333.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900004531.mm.1 | NR_045333.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900004531.mm.1 | TC0900000633.mm.1 | MTA TC |
