Assay Details
| Gene Symbol: Gm9484 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK142996.1 | - | 1 | 751 | 62 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 8013 | Gm8013 | EG666266 | -61.3kb | Anti-sense | Chr. 5 - 96911764 - 96934867 on NCBI Build GRCm38 |
| Downstream | BMP2 inducible kinase | Bmp2k |
4933417M22Rik, AA673486, AV128808, BIKE |
325.0 | Anti-sense | Chr. 5 - 96997689 - 97091048 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm9484
Genomic Map