Assay Details
| Gene Symbol: Gm6300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033591.2 | 1 - 2 | 755 | 109 | ||
| GenBank mRNA | AK143764.1 | - | 1 - 2 | 758 | 109 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RALY RNA binding protein-like | Ralyl | 0710005M24Rik | -180.8kb | Sense | Chr. 3 - 13471655 - 14182287 on NCBI Build GRCm38 |
| Downstream |
solute carrier family 7 (cationic amino acid transporter, y+ system), member 12 |
Slc7a12 |
Asc-2, XAT1 |
109.8kb | Sense | Chr. 3 - 14480691 - 14505819 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm6300
Genomic Map