Assay Details
Gene Symbol: Gm12718 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040673.1 | 1 - 2 | 98 | 76 | ||
GenBank mRNA | AK144477.1 | - | 1 - 2 | 99 | 76 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
OMA1 homolog, zinc metallopeptidase (S. cerevisiae) |
Oma1 |
2010001O09Rik, MPRP-1, ZMPOMA1 |
-16.1kb | Sense | Chr. 4 - 103313209 - 103366438 on NCBI Build GRCm38 |
Downstream | disabled 1 | Dab1 |
AI956902, C630028C02Rik, scm, scr, scrambler, yot |
214.8kb | Sense | Chr. 4 - 103619546 - 104744844 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 408420 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 430174 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 503346 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 135138 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 11817 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 588164 | AB093290.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 588164 | AK034089.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 588164 | XM_132047.7 | refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 588164 | XM_914264.3 | refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 728237 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 506967 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 579069 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 367388 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 695611 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 423923 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 213164 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 469211 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 263360 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 336502 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 25222 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 930874 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 177420 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 578286 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 79176 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 284409 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 307710 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 511551 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 951089 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 546681 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 508907 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 935699 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 106108 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 585444 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 715679 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 125335 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 409187 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 955150 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 586234 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 796725 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 969129 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 158148 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 309279 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 432854 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 241086 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 121831 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 117412 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 166988 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 84964 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 54114 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 29187 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P128769 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05341260 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P128769 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00947032 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00947046 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00947177 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05341291 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05341344 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05341366 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05341317 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05341291 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P00947177 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P128769 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03297662 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23165858 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23165842 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23165883 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23166436 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23165911 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03425603 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03377350 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23166439 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23165835 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23166079 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23166443 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23165826 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03511506 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04009745 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03696244 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23165995 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23166080 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23165913 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03297673 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03892381 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03952751 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23165828 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23165844 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03518303 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03932837 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03963298 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23165836 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03696248 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03243987 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23165832 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23165997 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23166008 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23166003 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23166020 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23166096 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23166022 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23166094 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23166076 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23166089 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23166445 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23165847 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23165853 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23166023 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23165963 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23166087 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23166430 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03672194 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23165891 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23165996 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23165870 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03344727 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23165912 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23166027 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23165852 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04009747 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23165833 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03963310 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23166070 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23165907 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23166441 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23166093 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23166000 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23165862 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23166090 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23166005 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23165865 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23166431 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03696249 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03260639 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23166012 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23165880 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23165968 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23166073 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03243990 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23166007 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23165838 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23165882 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03515352 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23165866 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23165849 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23165956 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23166444 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03892382 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03940757 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23165906 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03618366 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23165885 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23165894 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23165878 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23165843 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23165868 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23165954 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23165840 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23165859 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23166438 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03696247 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23166013 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23165888 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23166015 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23166433 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23166075 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23166083 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23165964 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23165889 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04067855 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23165845 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23166006 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Clariom S Array | PSR0400007346.mm.1 | NR_040673.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400007346.mm.1 | NR_040673.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400007346.mm.1 | TC0400001073.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400007350.mm.1 | NR_040673.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400007350.mm.1 | NR_040673.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400007350.mm.1 | TC0400001073.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400007352.mm.1 | NR_040673.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400007352.mm.1 | NR_040673.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400007352.mm.1 | TC0400001073.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400007348.mm.1 | NR_040673.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400007348.mm.1 | NR_040673.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400007348.mm.1 | TC0400001073.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400007347.mm.1 | NR_040673.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400007347.mm.1 | NR_040673.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400007347.mm.1 | TC0400001073.mm.1 | MTA TC |
