Assay Details
| Gene Symbol: 9330188P03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK145081.1 | - | 119 | 93 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 35909 |
Gm35909 | -66.1kb | Sense | Chr. 14 - 105518096 - 105523485 on NCBI Build GRCm38 | |
| Downstream | microRNA 6390 | Mir6390 |
Gm27562, mmu-mir-6390 |
32.0kb | Anti-sense | Chr. 14 - 105623642 - 105623770 on NCBI Build GRCm38 |
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 203126 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 350234 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 419228 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 825500 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 720218 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2419021 | AK034417.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08227336 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29877931 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29877891 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29878041 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11159752 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29877987 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29877997 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11485901 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29877998 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29877916 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11209779 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29878024 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29877967 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11464565 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29877994 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29877936 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29877985 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29877903 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29878029 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29878003 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29877910 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29877917 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29877908 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29877899 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29878009 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29878044 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29877904 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29877913 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11816152 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29877930 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29877992 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29877934 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29878026 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29877990 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29877901 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29877982 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11279147 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29878048 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29878020 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29878005 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29877889 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29877895 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29877969 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29878050 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29878021 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11579515 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29877914 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11279148 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29878014 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29878012 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29877977 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29877888 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11386832 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29877991 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11458349 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29877911 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29878034 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29878007 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29878052 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29877896 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29877929 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29877892 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29877918 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29878028 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29877912 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29878046 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29877932 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29878033 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29877980 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29878025 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29878049 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29877886 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29878010 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11464571 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29877937 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11209774 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29877884 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11555022 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29877971 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29877906 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29878001 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29877924 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29878045 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29877972 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 800895 | - | - |

9330188P03Rik
Genomic Map