Assay Details
| Gene Symbol: Gm29685 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK147773.1 | - | 400 | 74 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | otogelin-like | Otogl |
EG628870, Gm6924 |
-56.0kb | Anti-sense | Chr. 10 - 107760530 - 107912174 on NCBI Build GRCm38 |
| Downstream |
protein phosphatase 1, regulatory (inhibitor) subunit 12A |
Ppp1r12a |
1200015F06Rik, 5730577I22Rik, AA792106, AV099298, D10Ertd625e, Mypt1 |
192.8kb | Sense | Chr. 10 - 108162065 - 108279646 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm29685
Genomic Map