Assay Details
| Gene Symbol: Gm10067 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK147851.1 | - | 2 - 3 | 324 | 72 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
family with sequence similarity 155, member A |
Fam155a |
6430500D16, AW121567, Tmem28 |
-108.0kb | Sense | Chr. 8 - 9206010 - 9773363 on NCBI Build GRCm38 |
| Downstream |
ligase IV, DNA, ATP-dependent |
Lig4 |
5830471N16Rik, tiny |
53.3kb | Sense | Chr. 8 - 9970020 - 9976323 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK147851.1 transcripts.
Set Membership: |
|

Gm10067
Genomic Map