Assay Details
Gene Symbol: 4930513N10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_015574.2 | 1 - 2 | 2346 | 72 | ||
GenBank mRNA | AK147860.1 | - | 1 - 2 | 2347 | 72 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA 7073 | Mir7073 | mmu-mir-7073 | -53.6kb | Anti-sense | Chr. 8 - 95753151 - 95753214 on NCBI Build GRCm38 |
Downstream | predicted gene 10094 | Gm10094 | ENSMUSG00000061104 | 15.1kb | Sense | Chr. 8 - 95825356 - 95826156 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P499299 | AK161453.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P499299 | AK042481.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P499299 | AK029729.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P499299 | AK161453.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P499299 | AK042481.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P499299 | AK029729.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 603817 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 688335 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 618012 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 741779 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 180997 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 676880 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 353077 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 649116 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 126309 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P499299 | AK161453.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P499299 | AK042481.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P499299 | AK029729.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P499299 | AK161453.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P499299 | AK042481.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P499299 | AK029729.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01803621 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06606380 | - | - |
Whole Mouse Genome G4122A | A_52_P499299 | AK161453.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P499299 | AK042481.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P499299 | AK029729.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P499299 | AK161453.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P499299 | AK042481.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P499299 | AK029729.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111952 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26085440 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26085451 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07044178 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07273063 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26085408 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26085394 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26085388 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07324050 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07324048 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06851972 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06851405 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26085438 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26085400 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07234595 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07097633 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06771427 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26085411 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26085392 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07125510 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07208594 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26085377 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26085406 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26085398 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26085405 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26085450 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26085412 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07099097 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26085399 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26085410 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26085396 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26085383 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26085448 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26085434 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26085386 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26085393 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07234598 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07179448 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07086974 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07234594 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07324044 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26085404 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07099101 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26085442 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26085385 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT MG-430 PM Array Plate | 1457415_PM_a_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1442152_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1441278_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10574410 | - | - |
Mouse Genome 430 2.0 Array | 1441278_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1441278_at | - | - |
Mouse Genome 430 2.0 Array | 1442152_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1457415_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1457415_a_at | - | - |
Mouse Genome 430 2.0 Array | 1442152_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0800009052.mm.1 | NR_015574.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800009052.mm.1 | TC0800001146.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800009045.mm.1 | NR_015574.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800009045.mm.1 | TC0800001146.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800009051.mm.1 | NR_015574.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800009051.mm.1 | TC0800001146.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800009047.mm.1 | NR_015574.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800009047.mm.1 | TC0800001146.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800009050.mm.1 | NR_015574.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800009050.mm.1 | TC0800001146.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800009046.mm.1 | NR_015574.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800009046.mm.1 | TC0800001146.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800009049.mm.1 | NR_015574.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800009049.mm.1 | TC0800001146.mm.1 | MTA TC |
Mouse Expression Set 430 | 1457415_PM_a_at | - | - |
Mouse Expression Set 430 | 1442152_PM_at | - | - |
Mouse Expression Set 430 | 1457415_a_at | - | - |
Mouse Expression Set 430 | 1441278_PM_at | - | - |
Mouse Expression Set 430 | 1442152_at | - | - |
Mouse Expression Set 430 | 1441278_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1442152_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1457415_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1442152_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1441278_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1441278_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1457415_a_at | - | - |
