Assay Details
| Gene Symbol: Lpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK080872.1 | - | 1 | 1050 | 87 | |
| AK154419.1 | - | 1 | 1047 | 87 | ||
| AK170106.1 | - | 1 | 878 | 87 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | integrator complex subunit 10 | Ints10 |
4921521J11Rik, AI462004 |
-51.1kb | Sense | Chr. 8 - 68793928 - 68829410 on NCBI Build GRCm38 |
| Downstream |
solute carrier family 18 (vesicular monoamine), member 1 |
Slc18a1 |
4832416I10Rik, Vat1, Vmat1 |
156.0kb | Anti-sense | Chr. 8 - 69037708 - 69090629 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Lpl
Genomic Map