Assay Details
Gene Symbol: Tctn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK155612.1 | - | 114 | 117 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
aldehyde dehydrogenase 18 family, member A1 |
Aldh18a1 |
2810433K04Rik, AI429789, Pycs |
-972.0 | Anti-sense | Chr. 19 - 40550257 - 40588463 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
