Assay Details
| Gene Symbol: Arhgdia | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133796.7 | NP_598557.3 | 3 - 4 | 402 | 103 | |
| GenBank mRNA | AK075656.1 | - | 2 - 3 | 269 | 103 | |
| AK077343.1 | - | 3 - 4 | 562 | 103 | ||
| AK143516.1 | - | 3 - 4 | 386 | 103 | ||
| AK147842.1 | - | 361 | 103 | |||
| AK156819.1 | - | 347 | 103 | |||
| AK165241.1 | - | 3 - 4 | 384 | 103 | ||
| AK170553.1 | - | 3 - 4 | 369 | 103 | ||
| AK170718.1 | - | 3 - 4 | 386 | 103 | ||
| AK172140.1 | - | 347 | 103 | |||
| BC004732.1 | - | 3 - 4 | 344 | 3583026 | 103 | |
| BC086755.1 | - | 3 - 4 | 326 | 6414768 | 103 | |
More Information
Set Membership: |
|

Arhgdia
Genomic Map