Assay Details
Gene Symbol: AY702103 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK161492.1 | - | 247 | 83 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | raftlin lipid raft linker 1 | Rftn1 |
2310015N21Rik, AI853526 |
-49.2kb | Anti-sense | Chr. 17 - 49993258 - 50190497 on NCBI Build GRCm38 |
Downstream | deleted in azoospermia-like | Dazl |
Daz-like, Dazh, Dazl1, Dazla, Tpx-2, Tpx2 |
39.1kb | Anti-sense | Chr. 17 - 50279393 - 50293620 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
