Assay Details
Gene Symbol: Klhdc8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK161517.1 | - | 1 - 2 | 574 | 72 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | cDNA sequence BC048562 | BC048562 | -13.1kb | Anti-sense | Chr. 9 - 108436482 - 108446083 on NCBI Build GRCm38 | |
Downstream |
laminin, beta 2 |
Lamb2 |
AW211941, Lamb-2, Lams, npht |
19.8kb | Anti-sense | Chr. 9 - 108479857 - 108490530 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
