Assay Details
| Gene Symbol: Hhat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK163268.1 | - | 3 - 4 | 254 | 87 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
potassium voltage-gated channel, subfamily H (eag-related), member 1 |
Kcnh1 |
EAG1, Kv10.1, M-eag |
-144.8kb | Anti-sense | Chr. 1 - 192190362 - 192510159 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA A730013G03 gene | A730013G03Rik | 171.7kb | Sense | Chr. 1 - 192833173 - 192835808 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Hhat
Genomic Map