Assay Details
Gene Symbol: Vcpip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK079052.1 | - | 1 | 1630 | 86 | |
AK163653.1 | - | 1 | 45 | 86 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | myeloblastosis oncogene-like 1 | Mybl1 |
A-myb, G1-419-6, repro9 |
-20.1kb | Sense | Chr. 1 - 9667407 - 9700439 on NCBI Build GRCm38 |
Downstream | RIKEN cDNA 1700034P13 gene | 1700034P13Rik | 24.9kb | Anti-sense | Chr. 1 - 9747648 - 9771256 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
