Assay Details
| Gene Symbol: Mcu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK166044.1 | - | 3 - 4 | 405 | 70 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | oncoprotein induced transcript 3 | Oit3 |
EF-9, LZP |
-91.1kb | Sense | Chr. 10 - 59422960 - 59441779 on NCBI Build GRCm38 |
| Downstream | mitochondrial calcium uptake 1 | Micu1 |
C730016L05Rik, Calc, Cbara1 |
102.4kb | Anti-sense | Chr. 10 - 59702482 - 59864134 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Mcu
Genomic Map