Assay Details
| Gene Symbol: Idi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177197.4 | NP_796171.1 | 3 - 4 | 289 | 73 | |
| GenBank mRNA | AK029364.1 | - | 3 - 4 | 290 | 73 | |
| AK081319.1 | - | 3 - 4 | 290 | 73 | ||
| AY263178.1 | - | 3 - 4 | 290 | 73 | ||
| BC099586.1 | - | 3 - 4 | 276 | 30292324 | 73 | |
| BC125310.1 | - | 3 - 4 | 275 | 40129825 | 73 | |
| BC138100.1 | - | 3 - 4 | 275 | 8861119 | 73 | |
More Information
Set Membership: |
|

Idi2
Genomic Map