Assay Details
Gene Symbol: AY512915 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033559.1 | 2 - 3 | 261 | 93 | ||
GenBank mRNA | AK139655.1 | - | 2 - 3 | 262 | 93 | |
AY512915.1 | - | 246 | 93 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | kelch repeat and BTB (POZ) domain containing 8 | Kbtbd8 |
AU040152, SSEC-51, SSEC51, Takrp, mKIAA1842 |
-168.0kb | Anti-sense | Chr. 6 - 95117880 - 95129793 on NCBI Build GRCm38 |
Downstream |
succinate-Coenzyme A ligase, GDP-forming, beta subunit |
Suclg2 |
AF171077, AW556404, D6Wsu120e, SCS-betaG |
145.5kb | Sense | Chr. 6 - 95473009 - 95718846 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P101125 | AY512915.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2232729 | AK139655.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2232729 | AU045440.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2232729 | NR_033559.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P130261 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P169974 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2232729 | AK139655.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2232729 | AU045440.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2232729 | NR_033559.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P101125 | AY512915.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01390985 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01390970 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P130261 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05990354 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P169974 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101125 | AY512915.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P130261 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01390985 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P169974 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661450 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05346545 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661520 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05346541 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05497065 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661625 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661526 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05845240 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05845239 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661669 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05741868 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661600 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05359771 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05319228 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661516 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661534 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05346543 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661829 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661835 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661558 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661597 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661788 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661462 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661776 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661621 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661540 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661456 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661627 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661515 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05845243 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05822042 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05451254 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05497076 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661767 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661654 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661662 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661522 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661463 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661659 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661805 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05346546 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661454 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05346544 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661764 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661811 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661831 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05319239 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661470 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661564 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661466 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661712 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661573 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05710791 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05752147 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05382822 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661741 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661773 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05508660 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661798 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661452 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661824 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661785 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661518 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661836 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05497073 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661620 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05058467 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661739 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661598 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661717 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661814 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661807 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661656 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661710 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661797 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661819 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661512 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661809 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661778 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661769 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661763 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05767420 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661605 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661749 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661624 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661768 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661513 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05372239 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661533 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661455 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661560 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05359766 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661821 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661795 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661532 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05346548 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05627302 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661791 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661524 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661830 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661458 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661803 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661572 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661581 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05497067 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661468 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661570 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05640356 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05204551 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05822048 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661660 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661667 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661576 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661780 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661622 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05767418 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661557 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661562 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661796 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05458713 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661786 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661535 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661655 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661652 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661464 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661757 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661469 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05346552 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661460 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661766 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661834 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05044013 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661606 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661579 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05723316 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661707 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661541 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661566 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05752146 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05779897 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661465 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661745 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661804 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661800 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661738 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24661568 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05627308 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05047885 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05346547 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT MG-430 PM Array Plate | 1445526_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10546535 | - | - |
Mouse Genome 430 2.0 Array | 1445526_at | - | - |
Mouse Genome 430 2.0 Array | 1445526_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0600021579.mm.1 | TC0600002782.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600021579.mm.1 | NR_033559.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600021579.mm.1 | NR_033559.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600021576.mm.1 | TC0600002782.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600021576.mm.1 | NR_033559.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600021576.mm.1 | NR_033559.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600021575.mm.1 | TC0600002782.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600021575.mm.1 | NR_033559.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600021575.mm.1 | NR_033559.1 | Refseq |
Mouse Expression Set 430 | 1445526_PM_at | - | - |
Mouse Expression Set 430 | 1445526_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1445526_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1445526_at | - | - |
