Assay Details
Gene Symbol: 2210018M11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK046354.1 | - | 1 | 1099 | 62 | |
BC023871.1 | - | 1 | 843 | 62 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | leucine rich repeat containing 32 | Lrrc32 |
AI426318, D11S833Eh, D7H11S833E, EG434215, Garp |
-87.7kb | Anti-sense | Chr. 7 - 98489301 - 98501831 on NCBI Build GRCm38 |
Downstream | predicted gene 15506 | Gm15506 | OTTMUSG00000023003 | 91.8kb | Sense | Chr. 7 - 98682331 - 98703391 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
