Assay Details
| Gene Symbol: BC037156 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC037156.1 | - | 3 - 4 | 147 | 66 | |
| JQ408704.1 | - | 3 - 4 | 137 | 66 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 1700066N21 gene | 1700066N21Rik | Anti-sense | Chr. 5 - 87908588 - 87979351 on NCBI Build GRCm38 | ||
| Downstream | casein kappa | Csn3 |
AW208918, CSN10, Csnk |
12.5kb | Sense | Chr. 5 - 87925575 - 87932545 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

BC037156
Genomic Map