Assay Details
| Gene Symbol: Lppos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037954.1 | 2 | 758 | 83 | ||
| GenBank mRNA | AK004252.1 | - | 2 | 546 | 83 | |
| AK039760.1 | - | 564 | 83 | |||
| BC037367.1 | - | 1 | 452 | 83 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 38456 |
Gm38456 | -96.3kb | Sense | Chr. 16 - 24271143 - 24296233 on NCBI Build GRCm38 | |
| Downstream | LIM domain containing preferred translocation partner in lipoma | Lpp |
9430020K16Rik, AA959454, AU024130, B130055L10Rik, C79715, D630048H16 |
247.0 | Anti-sense | Chr. 16 - 24393350 - 24992578 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Lppos
Genomic Map