Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700017489.mm.1 |
NR_045146.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017489.mm.1 |
NR_045146.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017489.mm.1 |
TC1700001884.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700017494.mm.1 |
NR_045146.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017494.mm.1 |
NR_045146.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017494.mm.1 |
TC1700001884.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700017488.mm.1 |
NR_045146.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017488.mm.1 |
NR_045146.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017488.mm.1 |
TC1700001884.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700017490.mm.1 |
NR_045146.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017490.mm.1 |
NR_045146.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017490.mm.1 |
TC1700001884.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700017492.mm.1 |
NR_045146.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017492.mm.1 |
NR_045146.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017492.mm.1 |
TC1700001884.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700017493.mm.1 |
NR_045146.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017493.mm.1 |
NR_045146.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017493.mm.1 |
TC1700001884.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700017491.mm.1 |
NR_045146.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017491.mm.1 |
NR_045146.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700017491.mm.1 |
TC1700001884.mm.1 |
MTA TC |