Assay Details
| Gene Symbol: BC058420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC052319.1 | - | 1 | 2850 | 84 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | G protein-coupled receptor 26 | Gpr26 |
9630036A11Rik, A730021D22 |
-2.7kb | Sense | Chr. 7 - 131966135 - 131985633 on NCBI Build GRCm38 |
| Downstream | carboxypeptidase X 2 (M14 family) | Cpxm2 |
4632435C11Rik, CPX-2, Cpx2 |
38.3kb | Anti-sense | Chr. 7 - 132032687 - 132154768 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

BC058420
Genomic Map