Assay Details
| Gene Symbol: Gm2848 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046069.1 | 2 - 3 | 237 | 57 | ||
| GenBank mRNA | BC065398.1 | - | 2 - 3 | 237 | 57 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
DIRAS family, GTP-binding RAS-like 2 |
Diras2 |
2900052J15Rik, AI414999 |
-39.2kb | Sense | Chr. 13 - 52504375 - 52531242 on NCBI Build GRCm38 |
| Downstream | spleen tyrosine kinase | Syk | Sykb | 11.5kb | Anti-sense | Chr. 13 - 52582329 - 52648792 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC1700000218.rn
Set Membership: |
|

Gm2848
Genomic Map