Assay Details
Gene Symbol: Snhg14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC070450.1 | - | 1288 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2207922 | BC070450.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 137761 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608195 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 432538 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 31414 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 116564 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 441408 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 595538 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 336498 | AK005650.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 757040 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 313159 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 438434 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 577066 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 812497 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 139675 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 190478 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 328499 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 814673 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 674468 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 366280 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 175334 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 56891 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 153322 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2207922 | BC070450.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01527171 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06211321 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06211349 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06211324 | - | - |
Whole Mouse Genome G4122A | A_52_P145433 | AK035645.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P145433 | AK078094.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P145433 | AK035508.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P145433 | NR_015456.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109411 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06211349 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25159119 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25159110 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25159118 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25159112 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25159115 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25159127 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25159097 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25159133 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25159124 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25159126 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25159107 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25159111 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25159123 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25159130 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25159129 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25159137 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25159098 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25159113 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25159108 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25159122 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25159102 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25159128 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25159095 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25159132 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25159134 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25159106 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25159121 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25159101 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25159136 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT MG-430 PM Array Plate | 1455087_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1445296_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1436964_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1445296_at | - | - |
Mouse Genome 430 2.0 Array | 1436964_at | - | - |
Mouse Genome 430 2.0 Array | 1455087_at | - | - |
Mouse Genome 430 2.0 Array | 1445296_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1436964_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1455087_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0700028271.mm.1 | NR_015479.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028271.mm.1 | NR_015479.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028271.mm.1 | TC0700003126.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700028669.mm.1 | NR_015456.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028669.mm.1 | NR_015456.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028669.mm.1 | TC0700003345.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700028677.mm.1 | NR_015456.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028677.mm.1 | NR_015456.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028677.mm.1 | TC0700003345.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700028675.mm.1 | NR_015456.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028675.mm.1 | NR_015456.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028675.mm.1 | TC0700003345.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700028265.mm.1 | NR_015479.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028265.mm.1 | NR_015479.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028265.mm.1 | TC0700003126.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700028676.mm.1 | NR_015456.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028676.mm.1 | NR_015456.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028676.mm.1 | TC0700003345.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700028680.mm.1 | NR_015456.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028680.mm.1 | NR_015456.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028680.mm.1 | TC0700003345.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700028270.mm.1 | NR_015479.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028270.mm.1 | NR_015479.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028270.mm.1 | TC0700003126.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700028674.mm.1 | NR_015456.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028674.mm.1 | NR_015456.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028674.mm.1 | TC0700003345.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700028666.mm.1 | NR_015456.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028666.mm.1 | NR_015456.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028666.mm.1 | TC0700003345.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700028682.mm.1 | NR_015456.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028682.mm.1 | NR_015456.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028682.mm.1 | TC0700003345.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700028668.mm.1 | NR_015456.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028668.mm.1 | NR_015456.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028668.mm.1 | TC0700003345.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700028670.mm.1 | NR_015456.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028670.mm.1 | NR_015456.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028670.mm.1 | TC0700003345.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700028671.mm.1 | NR_015456.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028671.mm.1 | NR_015456.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028671.mm.1 | TC0700003345.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700028264.mm.1 | NR_015479.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028264.mm.1 | NR_015479.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028264.mm.1 | TC0700003126.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700028679.mm.1 | NR_015456.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028679.mm.1 | NR_015456.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028679.mm.1 | TC0700003345.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700028667.mm.1 | NR_015456.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028667.mm.1 | NR_015456.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028667.mm.1 | TC0700003345.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700028683.mm.1 | NR_015456.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028683.mm.1 | NR_015456.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028683.mm.1 | TC0700003345.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700028267.mm.1 | NR_015479.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028267.mm.1 | NR_015479.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028267.mm.1 | TC0700003126.mm.1 | MTA TC |
Mouse Expression Set 430 | 1436964_at | - | - |
Mouse Expression Set 430 | 1445296_at | - | - |
Mouse Expression Set 430 | 1445296_PM_at | - | - |
Mouse Expression Set 430 | 1455087_at | - | - |
Mouse Expression Set 430 | 1436964_PM_at | - | - |
Mouse Expression Set 430 | 1455087_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1436964_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1445296_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1436964_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1455087_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1455087_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1445296_PM_at | - | - |
