Assay Details
Gene Symbol: Snhg14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC070450.1 | - | 1367 | 92 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome |
---|

More Information
Set Membership: |
![]() ![]() ![]() |
