Assay Details
Gene Symbol: LOC545966 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC079901.1 | - | 3 - 4 | 579 | 86 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | E2F transcription factor 8 | E2f8 |
4432406C08Rik, AA410048 |
-27.0kb | Anti-sense | Chr. 7 - 48866429 - 48882142 on NCBI Build GRCm38 |
Downstream | neuron navigator 2 | Nav2 |
5330421F07Rik, AU067780, BQ960892, HELAD1, POMFIL2, RAINB1, RAINB2, Unc53H2, mKIAA3015 |
46.0kb | Sense | Chr. 7 - 48959066 - 49610088 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 9997 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 216366 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 846646 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 569095 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 174612 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 126926 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06190327 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01511987 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25108877 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06423998 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25108933 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25108964 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25108913 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25108909 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06383744 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25109042 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06439771 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25109041 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06097195 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25108931 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06276638 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05917861 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25108963 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25108942 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25108898 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25108875 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06241513 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25108940 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25109037 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06097194 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25108897 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25109012 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25109045 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06554758 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06423999 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06423996 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25109065 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25109052 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25109026 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25109013 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25108957 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06519294 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05917857 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06078857 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25109024 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05917855 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25108864 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25108911 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25108950 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25109017 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05940831 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25108923 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25109049 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25109059 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25108935 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06541343 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25108881 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06055187 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25108899 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25108929 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25108874 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05927630 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06319483 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25108965 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06541346 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06476779 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25108914 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25108920 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25108925 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25108895 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06109674 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06272954 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06272952 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25109038 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06554756 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06019060 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06055108 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25108888 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06325880 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25108943 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25108886 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25108865 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25108932 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25108949 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25109022 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06439767 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25109014 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25108939 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06634529 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06228719 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25108871 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06272947 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06519306 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06055109 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06437684 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25109063 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25109040 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25108952 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25109036 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06319485 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25108956 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06272945 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25108916 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25108878 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25108867 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06634528 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25108885 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06519307 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25108901 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06329761 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25108900 | - | - |
