Assay Details
Gene Symbol: Fam109a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC095929.1 | - | 128 | 75 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | SH2B adaptor protein 3 | Sh2b3 |
AI429800, Lnk |
-10.6kb | Anti-sense | Chr. 5 - 121815481 - 121837655 on NCBI Build GRCm38 |
Downstream | cut-like homeobox 2 | Cux2 |
1700051K22Rik, Cutl2, Cux-2, ENSMUSG00000072641 |
7.4kb | Anti-sense | Chr. 5 - 121857962 - 122050562 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
