Assay Details
| Gene Symbol: Klra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170851.1 | NP_001164322.1 | 7 - 8 | 938 | 103 | |
| GenBank mRNA | EF025059.1 | - | 7 - 8 | 772 | 103 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 6600 | Gm6600 |
287g15T7, C86350, EG625558, Zfp-like |
-420.1kb | Sense | Chr. 6 - 130728932 - 130799970 on NCBI Build GRCm38 |
| Downstream | mago-nashi homolog B (Drosophila) | Magohb |
2010012C16Rik, Mago, Magoh, Magoh-rs1 |
62.5kb | Sense | Chr. 6 - 131284388 - 131293244 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Klra2
Genomic Map