Assay Details
| Gene Symbol: Fbxw11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK010033.1 | - | 1 | 654 | 72 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | serine/threonine kinase 10 | Stk10 |
Gek1, Lok, mKIAA4026 |
-55.5kb | Sense | Chr. 11 - 32533266 - 32624595 on NCBI Build GRCm38 |
| Downstream | small integral membrane protein 23 | Smim23 | 1700008A04Rik | 139.5kb | Anti-sense | Chr. 11 - 32820376 - 32824594 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Fbxw11
Genomic Map