Assay Details
| Gene Symbol: 2310074N15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK010155.1 | - | 1 | 450 | 91 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane and ubiquitin-like domain containing 1 | Tmub1 |
2010004O20Rik, AB030183, Hops |
Anti-sense | Chr. 5 - 24445463 - 24448331 on NCBI Build GRCm38 | |
| Downstream |
ArfGAP with GTPase domain, ankyrin repeat and PH domain 3 |
Agap3 |
AGAP-3, AV028425, Centg3, Crag, Crag., MRIP-1, cnt-g3 |
3.7kb | Sense | Chr. 5 - 24452177 - 24502047 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2310074N15Rik
Genomic Map