Assay Details
| Gene Symbol: Zpbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK011513.1 | - | 1 | 751 | 94 | |
| AK137467.1 | - | 1 | 1050 | 94 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | IKAROS family zinc finger 3 | Ikzf3 |
5830411O07Rik, Aiolos, Zfpn1a3, Znfn1a3 |
-5.0kb | Anti-sense | Chr. 11 - 98464610 - 98546380 on NCBI Build GRCm38 |
| Downstream | ORM1-like 3 (S. cerevisiae) | Ormdl3 |
2810011N17Rik, AI315223 |
28.8kb | Anti-sense | Chr. 11 - 98581294 - 98587245 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Zpbp2
Genomic Map