Assay Details
| Gene Symbol: 2610300A13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK011940.1 | - | 1 | 252 | 94 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
AF4/FMR2 family, member 3 |
Aff3 |
3222402O04Rik, A730046J16, LAF-4, Laf4 |
-139.3kb | Anti-sense | Chr. 1 - 38175991 - 38627837 on NCBI Build GRCm38 |
| Downstream | LON peptidase N-terminal domain and ring finger 2 | Lonrf2 |
2900060P06Rik, AI851349 |
26.0kb | Anti-sense | Chr. 1 - 38793669 - 38836719 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2610300A13Rik
Genomic Map