Assay Details
Gene Symbol: 2700008L21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK012223.1 | - | 1 | 554 | 85 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | misshapen-like kinase 1 (zebrafish) | Mink1 |
B55, MINK, Map4k6, Ysk2 |
Sense | Chr. 11 - 70562594 - 70614482 on NCBI Build GRCm38 | |
Downstream |
cholinergic receptor, nicotinic, epsilon polypeptide |
Chrne |
AChrepsilon, Acre, nAChRE |
43.5kb | Anti-sense | Chr. 11 - 70614883 - 70619835 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
