Assay Details
| Gene Symbol: 2700054B07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK012420.1 | - | 1 | 151 | 87 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA E130308A19 gene | E130308A19Rik | -9.8kb | Anti-sense | Chr. 4 - 59626200 - 59759287 on NCBI Build GRCm38 | |
| Downstream | INTS3 and NABP interacting protein | Inip |
1110054O05Rik, 2610312O17Rik, AA399876, Ssbip1 |
251.0 | Sense | Chr. 4 - 59769642 - 59783855 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2700054B07Rik
Genomic Map