Assay Details
| Gene Symbol: 2810457G06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK013359.1 | - | 1 - 2 | 1104 | 105 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | sterile alpha motif domain containing 4 | Samd4 |
1700024G08Rik, 1700111L17Rik, 4933436G17Rik, Samd4a, Smaug, Smaug1 |
Anti-sense | Chr. 14 - 46882965 - 47105817 on NCBI Build GRCm38 | |
| Downstream | microRNA 378c | Mir378c |
Gm27930, mmu-mir-378c |
69.8kb | Sense | Chr. 14 - 46954830 - 46954928 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

2810457G06Rik
Genomic Map