Assay Details
| Gene Symbol: 2900006B11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK013488.1 | - | 1 | 1154 | 72 | |
| BM239197.2 | - | 1 | 72 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | hepatic leukemia factor | Hlf | E230015K02Rik | Sense | Chr. 11 - 90336535 - 90390917 on NCBI Build GRCm38 | |
| Downstream | syntaxin binding protein 4 | Stxbp4 |
6030470M02Rik, Synip |
118.7kb | Sense | Chr. 11 - 90476492 - 90638123 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2900006B11Rik
Genomic Map