Assay Details
| Gene Symbol: 2900057C01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK013713.1 | - | 1 | 656 | 70 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
cirrhosis, autosomal recessive 1A (human) |
Cirh1a |
Cirhin, Naic, Teg-292, Tex292 |
-51.7kb | Sense | Chr. 8 - 106893636 - 106923094 on NCBI Build GRCm38 |
| Downstream | predicted gene 16208 | Gm16208 | OTTMUSG00000031756 | 53.3kb | Anti-sense | Chr. 8 - 107029411 - 107031192 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2900057C01Rik
Genomic Map