Assay Details
| Gene Symbol: 2900060K15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK013735.1 | - | 1 | 1247 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | MACRO domain containing 2 | Macrod2 |
1110033L15Rik, 2610107G07Rik, 2900006F19Rik |
Sense | Chr. 2 - 140395356 - 142392966 on NCBI Build GRCm38 | |
| Downstream | kinesin family member 16B | Kif16b |
8430434E15Rik, AA623607, AI789011, C80253, C80902, mKIAA1590 |
1.4mb | Anti-sense | Chr. 2 - 142618334 - 142901595 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2900060K15Rik
Genomic Map