Assay Details
Gene Symbol: 2900073C17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK013771.1 | - | 1 | 1048 | 102 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | teneurin transmembrane protein 3 | Tenm3 |
2610100B16Rik, Odz1, Odz3, Ten-m3, mKIAA1455 |
Sense | Chr. 8 - 48225665 - 48935162 on NCBI Build GRCm38 | |
Downstream |
predicted gene, 19744 |
Gm19744 | 743.0 | Anti-sense | Chr. 8 - 48843885 - 48870497 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2089113 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2089113 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850581 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850621 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850482 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850614 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850480 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850476 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850492 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850563 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850514 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850599 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06748927 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850507 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850532 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850562 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850619 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06680704 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850564 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850602 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850516 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850556 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850486 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850528 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850543 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850534 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850594 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850479 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850524 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850612 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850536 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850616 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850490 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850546 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850622 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850511 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850553 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850489 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850544 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850593 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850600 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850579 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06673800 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850583 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850531 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850526 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850484 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850477 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850596 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07104869 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850520 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850508 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850519 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850530 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07277135 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850568 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850499 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850558 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850598 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850578 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850538 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850497 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850549 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850588 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850495 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850590 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850521 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850517 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850580 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850551 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850485 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850504 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850513 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06995533 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850608 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850502 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850540 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06895412 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850604 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850610 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850498 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850595 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850554 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850560 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25850506 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT MG-430 PM Array Plate | 1454301_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1454301_at | - | - |
Mouse Genome 430 2.0 Array | 1454301_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73020_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73020_PM_at | - | - |
Mouse Expression Set 430 | 1454301_PM_at | - | - |
Mouse Expression Set 430 | 1454301_at | - | - |
Murine Genome U74v2 Set | 139176_PM_at | - | - |
Murine Genome U74v2 Set | 139176_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1454301_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1454301_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 102940037 | - | - |
