Assay Details
| Gene Symbol: 2900084O13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK013822.1 | - | 1 | 1054 | 94 | |
| BM117709.2 | - | 1 | 94 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | neurexin I | Nrxn1 |
1700062G21Rik, 9330127H16Rik, A230068P09Rik, mKIAA0578 |
Sense | Chr. 17 - 90033644 - 91093100 on NCBI Build GRCm38 | |
| Downstream | adenylate cyclase activating polypeptide 1 | Adcyap1 | PACAP | 2.3mb | Anti-sense | Chr. 17 - 93198904 - 93206541 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2900084O13Rik
Genomic Map