Assay Details
| Gene Symbol: 2900092O11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK013844.1 | - | 1 | 550 | 86 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | dual specificity phosphatase 10 | Dusp10 |
2610306G15Rik, AI158871, MKP-5, MKP5 |
-367.3kb | Sense | Chr. 1 - 184034461 - 184075636 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 1700112H15 gene | 1700112H15Rik | 84.1kb | Anti-sense | Chr. 1 - 184527841 - 184557691 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2900092O11Rik
Genomic Map