Assay Details
Gene Symbol: 3010001F23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045451.1 | 3 - 4 | 470 | 77 | ||
GenBank mRNA | AK013876.1 | - | 3 - 4 | 473 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 641972 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 697804 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 253327 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 742536 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 364705 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 532018 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 124844 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 517281 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 548522 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 73802 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 581874 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 882985 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 572661 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 860340 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 189089 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 717823 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 445169 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 239745 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 865278 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 20939 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 73428 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 110233 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 27879 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P147151 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P147151 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09461317 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03778201 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03778198 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03778224 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P147151 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15020389 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32749228 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32749237 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32749234 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32749232 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32749235 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32749230 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14771152 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15056525 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32749238 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT MG-430 PM Array Plate | 1457010_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1457010_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1457010_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0X00009849.mm.1 | NR_045451.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00009849.mm.1 | NR_045451.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00009849.mm.1 | NR_045452.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00009849.mm.1 | NR_045452.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00009849.mm.1 | TC0X00001545.mm.1 | MTA TC |
Mouse Expression Set 430 | 1457010_PM_at | - | - |
Mouse Expression Set 430 | 1457010_at | - | - |
Murine Genome U74v2 Set | 139504_PM_at | - | - |
Murine Genome U74v2 Set | 139504_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1457010_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1457010_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 106770435 | - | - |
