Assay Details
Gene Symbol: 3110001N23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK013968.1 | - | 1 | 451 | 74 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
predicted gene, 29681 |
Gm29681 | -110.2kb | Sense | Chr. 5 - 131391600 - 131414766 on NCBI Build GRCm38 | |
Downstream | RIKEN cDNA 4930563F08 gene | 4930563F08Rik | 354.6kb | Anti-sense | Chr. 5 - 131880269 - 131883768 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
