Assay Details
Gene Symbol: 3110031N09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK014107.1 | - | 1 | 57 | 65 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
Yip1 domain family, member 7 |
Yipf7 |
2310016N21Rik, Yip1b |
-13.9kb | Sense | Chr. 5 - 69516670 - 69542647 on NCBI Build GRCm38 |
Downstream | GUF1 GTPase homolog (S. cerevisiae) | Guf1 |
4631409J12, AA407526, EF-4 |
26.0 | Anti-sense | Chr. 5 - 69556908 - 69573630 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
